Order Entry
United Kingdom
ContactUsLinkComponent
Anti-DNMT1 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 4G11-C7]
Anti-DNMT1 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 4G11-C7]
Catalog # ABNOMAB11779
Supplier:  Abnova
Anti-DNMT1 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 4G11-C7]
Catalog # ABNOMAB11779
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DNA (cytosine-5-)-methyltransferase 1
  • Antigen symbol:
    DNMT1
  • Clonality:
    Monoclonal
  • Clone:
    4G11-C7
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    1786
  • Antigen synonyms:
    CXXC9|HSN1E|DNMT|AIM|MCMT|ADCADN
  • Amino acid number:
    620 to 950
  • Storage buffer:
    PBS (0,05% BSA, 0,05% sodium azide)
  • Storage temperature:
    Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids of 620-950 of human DNMT1.
  • Purification:
    Protein G Purification
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant DNMT1.

Recommended Dilutions: Western Blot: 2-4 ?g/ml; Immunohistochemistry: 1-2 ?g/ml

Type: Primary
Antigen: DNMT1
Clonality: Monoclonal
Clone: 4G11-C7
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: