Order Entry
United Kingdom
ContactUsLinkComponent
Anti-ACP1 Mouse Monoclonal Antibody [clone: 2A15]
Anti-ACP1 Mouse Monoclonal Antibody [clone: 2A15]
Catalog # ABNOH00000052-M10
Supplier:  Abnova
Anti-ACP1 Mouse Monoclonal Antibody [clone: 2A15]
Catalog # ABNOH00000052-M10
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Acid Phosphatase 1
  • Antigen symbol:
    ACP1
  • Clonality:
    Monoclonal
  • Clone:
    2A15
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    52
  • Antigen synonyms:
    LMW-PTP|HAAP
  • Amino acid number:
    1 to 158
  • Storage buffer:
    1x PBS, pH 7,4
  • Sequence:
    MAEQATKSVLFVCLGNICRSPIAEAVFRKLVTDQNISENWVIDSGAVSDWNVGRSPDPRAVSCLRNHGIHTAHKARQITKEDFATFDYILCMDESNLRDLNRKSNQVKTCKAKIELLGSYDPQKQLIIEDPYYGNDSDFETVYQQCVRCCRAFLEKAH
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    ACP1 (AAH07422, 1 a.a. ~ 158 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against a full-length recombinant ACP1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: ACP1
Clonality: Monoclonal
Clone: 2A15
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: