Order Entry
United Kingdom
ContactUsLinkComponent
Anti-SPARC Rabbit Polyclonal Antibody
Anti-SPARC Rabbit Polyclonal Antibody
  ABNOPAB28625
 :  Abnova
Anti-SPARC Rabbit Polyclonal Antibody
  ABNOPAB28625
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Secreted Protein, Acidic, Cysteine-Rich
  • Antigen symbol:
    SPARC
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    6678
  • Antigen synonyms:
    Cysteine-Rich Protein|Secreted Protein Acidic And Rich In Cysteine|ON|Basement-Membrane Protein 40|Osteonectin|BM-40
  • Storage buffer:
    PBS, pH 7,2 (40% glycerol, 0,02% sodium azide)
  • Sequence:
    VLVTLYERDEDNNLLTEKQKLRVKKIHENEKRLEAGDHPVELLARDFEKNYNMYIFPVHWQFGQLDQHPIDGYLSHTELAPLRAPLIPMEHCTTRFFETCDLDNDKYIALDEWAGCFGIKQKDIDKDL
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids of human SPARC.
  • Purification:
    Antigen affinity purification
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody raised against recombinant SPARC.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Western Blot: 1:250-1:500

Type: Primary
Antigen: SPARC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human