Anti-FABP5 Mouse Polyclonal Antibody
ABNOH00002171-B03P
: Abnova
- Antibody type:Primary
- Antigen name:Fatty Acid Binding Protein 5
- Antigen symbol:FABP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:2171
- Antigen synonyms:EFABP|PA-FABP|E-FABP|PAFABP|KFABP
- Amino acid number:1 to 135
- Storage buffer:1x PBS, pH 7,4
- Sequence:MATVQQLEGRWRLVDSKGFDEYMKELGVGIALRKMGAMAKPDCIITCDGKNLTIKTESTLKTTQFSCTLGEKFEETTADGRITQTVCNFTDGALVQHQEWDGKESTITRKLKDGKLVVECVMNNVTCTRIYEKVE
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:FABP5 (AAH19385, 1 a.a. ~ 135 a.a) full-length human protein.
- Size:50 μg
- Pk:50 µg
Mouse polyclonal antibody raised against a full-length human FABP5 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: FABP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human