Anti-SIGLEC15 Rabbit Polyclonal Antibody
Catalog # ABNOPAB25806
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Sialic acid binding Ig-like lectin 15
- Antigen symbol:SIGLEC15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:284266
- Antigen synonyms:Siglec-15|SIGLEC-15|CD33L3|CD33 antigen-like 3|SIGLEC15|Sialic acid-binding Ig-like lectin 15|HsT1361
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 15 amino acids at N-terminus of human SIGLEC15.
- Purification:Peptide affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of SIGLEC15.
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry: 2.5 ?g/ml
Type: Primary
Antigen: SIGLEC15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat