Anti-NFKB1 Rabbit Polyclonal Antibody
ABNOPAB25044
: Abnova
- Antibody type:Primary
- Antigen name:Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1
- Antigen symbol:NFKB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:4790
- Antigen synonyms:NFkappaB|EBP-1|p50|NFKB-p50|NFKB-p105|NF-kB1|NF-kappaB|p105|KBF1|NF-kappa-B
- Modification:phospho-Ser907
- Storage buffer:PBS, pH 7,2 (0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding S907 of NFKB1.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against synthetic phosphopeptide of NFKB1.
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:200; Immunoprecipitation: 1:50-1:200
Type: Primary
Antigen: NFKB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human