Anti-MAP3K5 Rabbit Polyclonal Antibody
Catalog # ABNOPAB25031
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase Kinase Kinase 5
- Antigen symbol:MAP3K5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:4217
- Antigen synonyms:ASK1, ASK1, MEKK5, MAPKKK5, ASK1, Mitogen-activated protein kinase kinase kinase 5, Apoptosis signal-regulating kinase 1, ASK-1
- Modification:phospho-Ser83
- Storage buffer:PBS, pH 7,2 (0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding S83 of MAP3K5.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic phosphopeptide of MAP3K5.
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:200
Type: Primary
Antigen: MAP3K5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human