Anti-ETS1 Mouse Monoclonal Antibody [clone: 8A8]
ABNOMAB10639
: Abnova
- Antibody type:Primary
- Antigen name:V-Ets Avian Erythroblastosis Virus E2 Oncogene Homolog 1
- Antigen symbol:ETS1
- Clonality:Monoclonal
- Clone:8A8
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:2113
- Antigen synonyms:EWSR2|ETS-1|p54|c-ets-1
- Storage buffer:0,03% sodium azide
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human ETS1.
- Size:100 μl
- Pk:100 µl
Mouse monoclonal antibody raised against partial recombinant ETS1.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400
Type: Primary
Antigen: ETS1
Clonality: Monoclonal
Clone: 8A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human