Anti-LPAR2 Rat Polyclonal Antibody
ABNOPAB24995
: Abnova
- Antibody type:Primary
- Antigen name:lysophosphatidic acid receptor 2
- Antigen symbol:LPAR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:9170
- Antigen synonyms:LPA-2|EDG4|LPA2|EDG-4
- Storage buffer:PBS, pH 7,2 (0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to LPAR2.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against synthetic peptide of LPAR2.
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry: 1:50-1:200
Type: Primary
Antigen: LPAR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human