Anti-BRINP1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB24923
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:bone morphogenetic protein/retinoic acid inducible neural-specific 1
- Antigen symbol:BRINP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1620
- Antigen synonyms:DBCCR1|FAM5A|DBC1
- Storage buffer:PBS, pH 7,2 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to DBC1.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of DBC1.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: BRINP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human