Order Entry
United Kingdom
ContactUsLinkComponent
Anti-FAS Rabbit Polyclonal Antibody
Anti-FAS Rabbit Polyclonal Antibody
Catalog # ABNOPAB24829
Supplier:  Abnova
Anti-FAS Rabbit Polyclonal Antibody
Catalog # ABNOPAB24829
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fatty Acid Synthase
  • Antigen symbol:
    FAS
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    355
  • Antigen synonyms:
    FASTM|APO-1|ALPS1A|TNFRSF6|CD95|APT1|FAS1
  • Storage buffer:
    1x PBS, pH 7,2 (0,05% sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to FAS.
  • Purification:
    Antigen affinity purification
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of FAS.

Recommended Dilutions: Western Blot: 1:500-1000; Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: FAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human