Anti-TIAF1 Mouse Monoclonal Antibody [clone: 3B9]
ABNOH00009220-M04
: Abnova
- Antibody type:Primary
- Antigen name:TGFB1-Induced Anti-apoptotic Factor 1
- Antigen symbol:TIAF1
- Clonality:Monoclonal
- Clone:3B9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:9220
- Antigen synonyms:TIAF|TGFB1-induced anti-apoptotic factor 1|SPR210|12 kDa TGF-beta-1-induced antiapoptotic factor|MAJN
- Amino acid number:1 to 115
- Storage buffer:1x PBS, pH 7,4
- Sequence:MSSPSSPFREQSFLCAAGDAGEESRVQVLKNEVRRGSPVLLGWVEQAYADKCVCGPSAPPAPTPPSLSQRVMCNDLFKVNPFQLQQFRADPSTASLLLCPGGLDHKLNLRGKAWG
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:TIAF1 (NP_004731.2, 1 a.a. ~ 115 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:100 µg
- Pk:100 µg
Mouse monoclonal antibody raised against a full-length recombinant TIAF1.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: TIAF1
Clonality: Monoclonal
Clone: 3B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human