Order Entry
United Kingdom
ContactUsLinkComponent
Anti-PSAP Rabbit Polyclonal Antibody
Anti-PSAP Rabbit Polyclonal Antibody
Catalog # ABNOH00005660-D01
Supplier:  Abnova
Anti-PSAP Rabbit Polyclonal Antibody
Catalog # ABNOH00005660-D01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Prostate Specific Acid Phosphatase
  • Antigen symbol:
    PSAP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    5660
  • Antigen synonyms:
    SAP1|PAP|prostatic acid phosphotase|prostatic acid phosphatase|GLBA
  • Amino acid number:
    1 to 524
  • Storage buffer:
    No additive
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    PSAP (NP_002769.1, 1 a.a. ~ 524 a.a) full-length human protein.
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against a full-length human PSAP protein.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PSAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human