Order Entry
United Kingdom
ContactUsLinkComponent
Anti-RARG Mouse Polyclonal Antibody
Anti-RARG Mouse Polyclonal Antibody
  ABNOH00005916-B02P
 :  Abnova
Anti-RARG Mouse Polyclonal Antibody
  ABNOH00005916-B02P
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Retinoic Acid Receptor Gamma
  • Antigen symbol:
    RARG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    5916
  • Antigen synonyms:
    RARC|NR1B3
  • Amino acid number:
    1 to 454
  • Storage buffer:
    1x PBS, pH 7,4
  • Sequence:
    MATNKERLFAAGALGPGSGYPGAGFPFAFPGALRGSPPFEMLSPSFRGLGQPDLPKEMASLSVETQSTSSEEMVPSSPSPPPPPRVYKPCFVCNDKSSGYHYGVSSCEGCKGFFRRSIQKNMVYTCHRDKNCIINKVTRNRCQYCRLQKCFEVGMSKEAVRNDRNKKKKEVKEEGSPDSYELSPQLEELITKVSKAHQETFPSLCQLGKYTTNSSADHRVQLDLGLWDKFSELATKCIIKIVEFAKRLPGFTGLSIADQITLLKAACLDILMLRICTRYTPEQDTMTFSDGLTLNRTQMHNAGFGPLTDLVFAFAGQLLPLEMDDTETGLLSAICLICGDRMDLEEPEKVDKLQEPLLEALRLYARRRRPSQPYMFPRMLMKITDLRGISTKGAERAITLKMEIPGPMPPLIREMLENPEMFEDDSSQPGPHPNASSEDEVPGGQGKGGLKSPA
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    RARG (NP_000957.1, 1 a.a. ~ 454 a.a) full-length human protein.
  • Size:
    50 μg
  • Pk:
    50 µg

 

 

Mouse polyclonal antibody raised against a full-length human RARG protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: RARG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human