Anti-RARA Mouse Polyclonal Antibody
ABNOH00005914-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Retinoic Acid Receptor Alpha
- Antigen symbol:RARA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:5914
- Antigen synonyms:Retinoic acid receptor|nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form|NR1B1|alpha polypeptide|RAR|nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR short form
- Amino acid number:1 to 462
- Storage buffer:1x PBS, pH 7,4
- Sequence:MASNSSSCPTPGGGHLNGYPVPPYAFFFPPMLGGLSPPGALTTLQHQLPVSGYSTPSPATIETQSSSSEEIVPSPPSPPPLPRIYKPCFVCQDKSSGYHYGVSACEGCKGFFRRSIQKNMVYTCHRDKNCIINKVTRNRCQYCRLQKCFEVGMSKESVRNDRNKKKKEVPKPECSESYTLTPEVGELIEKVRKAHQETFPALCQLGKYTTNNSSEQRVSLDIDLWDKFSELSTKCIIKTVEFAKQLPGFTTLTIADQITLLKAACLDILILRICTRYTPEQDTMTFSDGLTLNRTQMHNAGFGPLTDLVFAFANQLLPLEMDDAETGLLSAICLICGDRQDLEQPDRVDMLQEPLLEALKVYVRKRRPSRPHMFPKMLMKITDLRSISAKGAERVITLKMEIPGSMPPLIQEMLENSGGLDTLSGQPGGGGRDGGGLAPPPGSCSPSLSPSSNRSSPATHSP
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:RARA (ABM84670.1, 1 a.a. ~ 462 a.a) full-length human protein.
- Size:50 μg
- Pk:50 µg
Mouse polyclonal antibody raised against a full-length human RARA protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: RARA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human