Anti-FADS3 Mouse Monoclonal Antibody [clone: 3D2]
ABNOH00003995-M07A
: Abnova
- Antibody type:Primary
- Antigen name:Fatty Acid Desaturase 3
- Antigen symbol:FADS3
- Clonality:Monoclonal
- Clone:3D2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites fluid
- Gene ID:3995
- Antigen synonyms:CYB5RP|LLCDL3
- Amino acid number:16 to 113
- Sequence:PGAPLPTFCWEQIRAHDQPGDKWLVIERRVYDISRWAQRHPGGSRLIGHHGAEDATDAFRAFHQDLNFVRKFLQPLLIGELAPEEPSQDGPLNAQLVE
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:FADS3 (NP_068373, 16 a.a. ~ 113 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:200 μl
- Pk:200 µl
Mouse monoclonal antibody raised against a partial recombinant FADS3.
Type: Primary
Antigen: FADS3
Clonality: Monoclonal
Clone: 3D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human