Order Entry
United Kingdom
ContactUsLinkComponent
Anti-FPR2 Mouse Polyclonal Antibody
Anti-FPR2 Mouse Polyclonal Antibody
Catalog # ABNOH00002358-B02P
Supplier:  Abnova
Anti-FPR2 Mouse Polyclonal Antibody
Catalog # ABNOH00002358-B02P
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Formyl Peptide Receptor 2
  • Antigen symbol:
    FPR2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    2358
  • Antigen synonyms:
    FPRH1|FPRH2|FMLPX|FPRL1|FPR2A|FMLP-R-II|HM63|LXA4R|ALXR
  • Amino acid number:
    1 to 351
  • Storage buffer:
    1x PBS, pH 7,4
  • Sequence:
    METNFSTPLNEYEEVSYESAGYTVLRILPLVVLGVTFVLGVLGNGLVIWVAGFRMTRTVTTICYLNLALADFSFTATLPFLIVSMAMGEKWPFGWFLCKLIHIVVDINLFGSVFLIGFIALDRCICVLHPVWAQNHRTVSLAMKVIVGPWILALVLTLPVFLFLTTVTIPNGDTYCTFNFASWGGTPEERLKVAITMLTARGIIRFVIGFSLPMSIVAICYGLIAAKIHKKGMIKSSRPLRVLTAVVASFFICWFPFQLVALLGTVWLKEMLFYGKYKIIDILVNPTSSLAFFNSCLNPMLYVFVGQDFRERLIHSLPTSLERALSEDSAPTNDTAANSASPPAETELQAM
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    FPR2 (NP_001005738.1, 1 a.a. ~ 351 a.a) full-length human protein.
  • Size:
    50 μg
  • Pk:
    50 µg

Specifications

About this item

Mouse polyclonal antibody raised against a full-length human FPR2 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: FPR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human