Anti-FABP4 Mouse Polyclonal Antibody
ABNOH00002167-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Fatty Acid Binding Protein 4
- Antigen symbol:FABP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:2167
- Antigen synonyms:ALBP|A-FABP|adipocyte-type fatty acid-binding protein|fatty acid-binding protein 4|AFABP|aP2|HEL-S-104
- Amino acid number:1 to 132
- Storage buffer:1x PBS, pH 7,4
- Sequence:MCDAFVGTWKLVSSENFDDYMKEVGVGFATRKVAGMAKPNMIISVNGDVITIKSESTFKNTEISFILGQEFDEVTADDRKVKSTITLDGGVLVHVQKWDGKSTTIKRKREDDKLVVECVMKGVTSTRVYERA
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:FABP4 (AAH03672, 1 a.a. ~ 132 a.a) full-length human protein.
- Size:50 μg
- Pk:50 µg
Mouse polyclonal antibody raised against a full-length human FABP4 protein.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: FABP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human