Anti-CA9 Rabbit Polyclonal Antibody
Catalog # ABNOH00000768-D01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Carbonic Anhydrase 9
- Antigen symbol:CA9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:768
- Antigen synonyms:CAIX|carbonic dehydratase|membrane antigen MN|carbonic anhydrase 9|RCC-associated protein G250|MN|carbonate dehydratase IX|CA-IX|RCC-associated antigen G250|P54/58N|pMW1|renal cell carcinoma-associated antigen G250
- Amino acid number:1 to 459
- Storage buffer:No additive
- Sequence:MAPLCPSPWLPLLIPAPAPGLTVQLLLSLLLLMPVHPQRLPRMQEDSPLGGGSSGEDDPLGEEDLPSEEDSPREEDPPGEEDLPGEEDLPGEEDLPEVKPKSEEEGSLKLEDLPTVEAPGDPQEPQNNAHRDKEGDDQSHWRYGGDPPWPRVSPACAGRFQSPVDIRPQLAAFCPALRPLELLGFQLPPLPELRLRNNGHSVQLTLPPGLEMALGPGREYRALQLHLHWGAAGRPGSEHTVEGHRFPAEIHVVHLSTAFARVDEALGRPGGLAVLAAFLEEGPEENSAYEQLLSRLEEIAEEGSETQVPGLDISALLPSDFSRYFQYEGSLTTPPCAQGVIWTVFNQTVMLSAKQLHTLSDTLWGPGDSRLQLNFRATQPLNGRVIEASFPAGVDSSPRAAEPVQLNSCLAAGDILALVFGLLFAVTSVAFLVQMRRQHRRGTKGGVSYRPAEVAETGA
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:CA9 (NP_001207.1, 1 a.a. ~ 459 a.a) full-length human protein.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against a full-length human CA9 protein.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: CA9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human