Anti-DsbA tag Monoclonal Antibody
ABNOMAB0048-M01
: Abnova
- Antibody type:Primary
- Antigen name:DsbA tag
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Amino acid number:51 to 150
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:DsbA (NP_458051, 51 a.a. ~ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against a partial recombinant DsbA.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: DsbA tag
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a
Reactivity: Human