Anti-FAS Rabbit Polyclonal Antibody
Catalog # ABNOPAB0843
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fatty Acid Synthase
- Antigen symbol:FAS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:355
- Antigen synonyms:FASTM|APO-1|ALPS1A|TNFRSF6|CD95|APT1|FAS1
- Storage buffer:PBS (0,2% gelatin, 0,1% sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human FAS.
- Size:200 μg
- Pk:200 µg
Specifications
About this item
Rabbit polyclonal antibody raised against a synthetic peptide of FAS.
Recommended Dilutions: Western Blot: 1:200
Type: Primary
Antigen: FAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human