Order Entry
United Kingdom
ContactUsLinkComponent
Anti-PPAP2C Rabbit Polyclonal Antibody
Anti-PPAP2C Rabbit Polyclonal Antibody
Catalog # ABNOPAB10122
Supplier:  Abnova
Anti-PPAP2C Rabbit Polyclonal Antibody
Catalog # ABNOPAB10122
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Phosphatidic Acid Phosphatase Type 2C
  • Antigen symbol:
    PPAP2C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    8612
  • Antigen synonyms:
    PAP2-gamma|PAP2-G|Lipid phosphate phosphohydrolase 2|Phosphatidate phosphohydrolase type 2c|PAP2c|Phosphatidic acid phosphatase 2c|PAP-2c
  • Storage buffer:
    PBS (0,08% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide of human PPAP2C.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of PPAP2C.

Recommended Dilutions: Western Blot: 10 to 15 ?g/ml

Type: Primary
Antigen: PPAP2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human