Anti-PPAP2B Rabbit Polyclonal Antibody
Catalog # ABNOPAB10123
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Phosphatidic Acid Phosphatase Type 2B
- Antigen symbol:PPAP2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:8613
- Antigen synonyms:PAP2B|VCIP|Dri42|LPP3
- Storage buffer:PBS (0,08% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide of human PPAP2B.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of PPAP2B.
Recommended Dilutions: Western Blot: 10 to 15 ?g/ml
Type: Primary
Antigen: PPAP2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human