Anti-RASD2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB13305
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:RASD family member 2
- Antigen symbol:RASD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:23551
- Antigen synonyms:Ras homolog enriched in striatum|MGC,4834|Rhes|TEM2
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus 15 amino acids of human RASD2.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of RASD2.
Recommended Dilutions: Western Blot: 1-2 ?g/ml
Type: Primary
Antigen: RASD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat