Order Entry
United Kingdom
ContactUsLinkComponent
Anti-GFAP Mouse Monoclonal Antibody [clone: SB61a]
Anti-GFAP Mouse Monoclonal Antibody [clone: SB61a]
Catalog # ABNOMAB3335
Supplier:  Abnova
Anti-GFAP Mouse Monoclonal Antibody [clone: SB61a]
Catalog # ABNOMAB3335
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Glial fibrillary acidic protein
  • Antigen symbol:
    GFAP
  • Clonality:
    Monoclonal
  • Clone:
    SB61a
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    2670
  • Antigen synonyms:
    FLJ45472|Intermediate filament protein.|Astrocyte|p-GFAP Ser38|GFAP phospho S38|GFAP|Glial Fibrillary Acidic Protein
  • Storage buffer:
    100 mM BBS, pH 8.0
  • Storage temperature:
    Store at 4 °C.
  • Immunogen:
    Recombinant protein corresponding to human GFAP.
  • Pk:
    500 µG

Specifications

About this item

Mouse monoclonal antibody raised against recombinant GFAP.

Recommended Dilutions: ELISA: 1 ?g

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: SB61a
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human