Order Entry
United Kingdom
ContactUsLinkComponent
Anti-AQP6 Rabbit Polyclonal Antibody
Anti-AQP6 Rabbit Polyclonal Antibody
Catalog # ABNOPAB21937
Supplier:  Abnova
Anti-AQP6 Rabbit Polyclonal Antibody
Catalog # ABNOPAB21937
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Aquaporin 6
  • Antigen symbol:
    AQP6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    363
  • Antigen synonyms:
    AQP2L|KID
  • Storage buffer:
    PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
  • Sequence:
    PDTKTLAQRLAILTGTVEVGTGAGAGAEPLKKESQPGSGAVEME
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids of human AQP6.
  • Purification:
    Antigen affinity purification
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against recombinant AQP6.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200

Type: Primary
Antigen: AQP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human