Anti-ELOVL6 Rabbit Polyclonal Antibody
Catalog # ABNOPAB13032
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Elongation of very long chain fatty acids-like 6
- Antigen symbol:ELOVL6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:79071
- Antigen synonyms:Fatty acyl-CoA elongase|FACE|Long-chain fatty-acyl elongase|Elongation of very long chain fatty acids protein 6|hELO2|ELOVL6|LCE
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus 16 amino acids of human ELOVL6.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of ELOVL6.
Recommended Dilutions: Western Blot: 1-2 ?g/ml
Type: Primary
Antigen: ELOVL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat