Order Entry
United Kingdom
ContactUsLinkComponent
Anti-NANOG Mouse Monoclonal Antibody [clone: 5A10]
Anti-NANOG Mouse Monoclonal Antibody [clone: 5A10]
Catalog # ABNOMAB2042
Supplier:  Abnova
Anti-NANOG Mouse Monoclonal Antibody [clone: 5A10]
Catalog # ABNOMAB2042
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Homeobox protein NANOG
  • Antigen symbol:
    NANOG
  • Clonality:
    Monoclonal
  • Clone:
    5A10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    79923
  • Antigen synonyms:
    ENK|ecat4|2410002E02Rik
  • Amino acid number:
    1 to 154
  • Storage buffer:
    PBS, pH 7,4 (0,1% sodium azide)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to amino acids 1-154 of human NANOG.
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant NANOG.

Type: Primary
Antigen: NANOG
Clonality: Monoclonal
Clone: 5A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse