Anti-FABP1 Mouse Monoclonal Antibody [clone: 2G4]
ABNOMAB2050
: Abnova
- Antibody type:Primary
- Antigen name:Fatty Acid Binding Protein 1
- Antigen symbol:FABP1
- Clonality:Monoclonal
- Clone:2G4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:2168
- Antigen synonyms:SCP|FABPL|p14|SCP.|L-FABP
- Amino acid number:1 to 127
- Storage buffer:PBS, pH 7,4 (0,1% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 1-127 of human FABP1.
- Size:100 μl
- Pk:100 µl
Mouse monoclonal antibody raised against partial recombinant FABP1.
Type: Primary
Antigen: FABP1
Clonality: Monoclonal
Clone: 2G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human