Anti-GABRB3 Goat Polyclonal Antibody
Catalog # ABNOPAB11457
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Gamma-aminobutyric acid (GABA) A Receptor beta 3
- Antigen symbol:GABRB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:2562
- Antigen synonyms:beta3|Cp1|Gamma aminobutyric acid A receptor beta 3,A230092K12Rik|AW049585|Gabrb-3
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-EKTAKAKNDRSK
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human GABRB3.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of GABRB3.
Recommended Dilutions: ELISA: 1:8000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-4 ?g/ml
Type: Primary
Antigen: GABRB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human