Anti-NANP Rabbit Polyclonal Antibody
ABNOPAB16305
: Abnova
- Antibody type:Primary
- Antigen name:N-acetylneuraminic acid phosphatase
- Antigen symbol:NANP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Hamster,Rabbit,Gorilla,Human,Dog,Rat,Mouse
- Cross adsorption:No
- Format:Liquid
- Gene ID:140838
- Antigen synonyms:HDHD4|C20orf147|dJ694B14.3
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to human NANP.
- Purification:Immunoaffinity purification
- Size:50 μg
- Pk:50 µg
Rabbit polyclonal antibody raised against synthetic peptide of NANP.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 10-15 ?g/ml
Type: Primary
Antigen: NANP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Gorilla, Hamster, Human, Mouse, Rabbit, Rat