Anti-GABBR1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB14531
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Gamma-Aminobutyric Acid (GABA) B Receptor, 1
- Antigen symbol:GABBR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:81657
- Antigen synonyms:GABABR1|GABBR1-3|GPRC3A|GB1|dJ271M21.1.2|dJ271M21.1.1
- Modification:phospho-Ser923
- Storage buffer:10 mM HEPES, 150 mM NaCl, pH 7,5 (50% glycerol, 10% BSA)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding S923 of rat Gabbr1.
- Purification:Affinity purification
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic phosphopeptide of Gabbr1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: GABBR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat