Anti-GABRA6 Rabbit Polyclonal Antibody
ABNOPAB14539
: Abnova
- Antibody type:Primary
- Antigen name:Gamma-Aminobutyric Acid (GABA) A Receptor, Alpha 6
- Antigen symbol:GABRA6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Serum
- Gene ID:29708
- Antigen synonyms:GABAAR alpha6|Gamma-aminobutyric acid receptor subunit alpha-6|GABA(A) receptor subunit alpha-6|GABRA6
- Storage buffer:serum
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to rat Gabra6.
- Size:50 μl
- Pk:50 Tests
Rabbit polyclonal antibody raised against synthetic peptide of Gabra6.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: GABRA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat