Order Entry
United Kingdom
ContactUsLinkComponent
Anti-HLA-DRA Mouse Monoclonal Antibody [clone: TAL 1B5]
Anti-HLA-DRA Mouse Monoclonal Antibody [clone: TAL 1B5]
Catalog # ABNOMAB7019
Supplier:  Abnova
Anti-HLA-DRA Mouse Monoclonal Antibody [clone: TAL 1B5]
Catalog # ABNOMAB7019
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Major histocompatibility Complex, class II, DR alpha
  • Antigen symbol:
    HLA-DRA
  • Clonality:
    Monoclonal
  • Clone:
    TAL 1B5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    3122
  • Antigen synonyms:
    MLRW|HLA-DRA1
  • Storage buffer:
    PBS
  • Storage temperature:
    Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Human bristol 8 separated alpha chain preparation
  • Size:
    100 μg
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against HLA-DRA.

Type: Primary
Antigen: HLA-DRA
Clonality: Monoclonal
Clone: TAL 1B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human