Anti-PCSK9 Mouse Monoclonal Antibody [clone: KS-2C8]
Catalog # ABNOMAB8228
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Proprotein convertase subtilisin/kexin type 9
- Antigen symbol:PCSK9
- Clonality:Monoclonal
- Clone:KS-2C8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:255738
- Antigen synonyms:PCSK9|Neural apoptosis-regulated convertase 1|NARC-1|NARC1|Proprotein convertase subtilisin/kexin type 9|Subtilisin/kexin-like protease PC9|Proprotein convertase PC9|PSEC0052
- Storage buffer:10 mM phosphate buffer, 150 mM NaCl, pH 7,5 (50% glycerol)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant His fusion protein corresponding to full length human PCSK9.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant PCSK9.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunoprecipitation: 1-2 ?g/sample)
Type: Primary
Antigen: PCSK9
Clonality: Monoclonal
Clone: KS-2C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human