Anti-SIGLEC12 Goat Polyclonal Antibody
Catalog # ABNOPAB6335
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Sialic acid-binding Ig-like lectin 12
- Antigen symbol:SIGLEC12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Cross adsorption:No
- Format:Liquid
- Gene ID:89858
- Antigen synonyms:mSiglec-E|Siglec9|SIGLECL1|Siglec5|Siglec12|Cd170|Siglecl1
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-EAIGYEYSEINIPK
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human SIGLEC12.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of SIGLEC12.
Recommended Dilutions: ELISA: 1:128000
Type: Primary
Antigen: SIGLEC12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: