Anti-SPARC Mouse Monoclonal Antibody [clone: ON1-1]
Catalog # ABNOMAB8284
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Secreted Protein, Acidic, Cysteine-Rich
- Antigen symbol:SPARC
- Clonality:Monoclonal
- Clone:ON1-1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Pig,Rabbit,Human,Bovine
- Western blot:Yes
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:282077
- Antigen synonyms:Cysteine-Rich Protein|Secreted Protein Acidic And Rich In Cysteine|ON|Basement-Membrane Protein 40|Osteonectin|BM-40
- Storage buffer:PBS (1%BSA, 0,1% sodium azide)
- Storage temperature:Store at 4 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Bovine SPARC.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse polyclonal antibody raised against SPARC.
Recommended Dilutions: ELISA: 1-10 ?g/ml; Western Blot: 1-10 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1-10 ?g/ml
Type: Primary
Antigen: SPARC
Clonality: Monoclonal
Clone: ON1-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Pig, Rabbit