Anti-HBcAg Mouse Monoclonal Antibody [clone: 14-E-11]
Catalog # ABNOMAB5404
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Hepatitis B Core Antigen
- Antigen symbol:HBcAg
- Clonality:Monoclonal
- Clone:14-E-11
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Virus
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Serum
- Antigen synonyms:Hepatitis B virus nucleocapsid particles|HepB
- Storage buffer:Serum
- Storage temperature:Store at 4 °C. For long term storage store at – 80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to hepatitis B virus core antigen core.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant hepatitis B virus core antigen
Recommended Dilutions: Western Blot: 0.2 ?g/ml
Type: Primary
Antigen: HBcAg
Clonality: Monoclonal
Clone: 14-E-11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Virus