Anti-GFP Polyclonal Antibody
ABNOPAB14930
: Abnova
- Antibody type:Primary
- Antigen name:Green Fluorescence Protein
- Antigen symbol:GFP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Form:Serum
- Antigen synonyms:GFPuv|eGFP|Green Fluorescent Protein
- Storage buffer:0,1% sodium azide
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified GFP.
- Size:1 mL
- Pk:1 ml
Sheep polyclonal antibody raised against native GFP.
Type: Primary
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: