Anti-RAI1 Rabbit Polyclonal Antibody
ABNOPAB5547
: Abnova
- Antibody type:Primary
- Antigen name:Retinoic Acid Induced 1
- Antigen symbol:RAI1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:10743
- Antigen synonyms:MGC12824|SMCR|DKFZP434A139|SMS|KIAA1820|retinoic acid induced 1|RAI1_HUMAN.
- Storage buffer:PBS, pH 7,2 (50% glycerol, 0,01% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to N-terminus of human RAI1.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of RAI1.
Type: Primary
Antigen: RAI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human