Anti-SIGLEC11 Rabbit Polyclonal Antibody
ABNOPAB16743
: Abnova
- Antibody type:Primary
- Antigen name:Sialic acid binding Ig-like lectin 11
- Antigen symbol:SIGLEC11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:114132
- Antigen synonyms:Sialic acid-binding lectin 11|Siglec11|Sialic acid-binding Ig-like lectin 11
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus 18 amino acids of human SIGLEC11.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of SIGLEC11.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: SIGLEC11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human