Anti-ADH1A Goat Polyclonal Antibody
ABNOPAB6725
: Abnova
- Antibody type:Primary
- Antigen name:Alcohol dehydrogenase 1A
- Antigen symbol:ADH1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:124|125|126
- Antigen synonyms:ADH1
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:STAGKVMKCKA
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human ADH1A/ADH1B/ADH1C.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Goat polyclonal antibody raised against synthetic peptide of ADH1A/ADH1B/ADH1C.
Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.3-1.5 ?g/ml
Type: Primary
Antigen: ADH1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human