Anti-FFAR4 Goat Polyclonal Antibody
ABNOPAB18699
: Abnova
- Antibody type:Primary
- Antigen name:Free fatty acid receptor 4
- Antigen symbol:FFAR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Epitope:Center
- Cross adsorption:No
- Format:Liquid
- Gene ID:338557
- Antigen synonyms:GPR120|BMIQ10|GT01|PGR4|GPR129|O3FAR1
- Storage buffer:Tris saline, pH 7,3 (0,02% sodium azide, 0,5% BSA)
- Sequence:C-YSESHQIRVSQQD
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids at internal region of human O3FAR1.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Goat polyclonal antibody raised against synthetic peptide of O3FAR1.
Recommended Dilutions: ELISA: 1:16000
Type: Primary
Antigen: FFAR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: