Anti-ELOVL1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB17711
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:ELOVL fatty acid elongase 1
- Antigen symbol:ELOVL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:64834
- Antigen synonyms:Ssc1|CGI-88
- Storage buffer:PBS, pH 7,4 (150mM NaCl, 0,02% sodium azide, 50% glycerol)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal of human ELOVL1.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of ELOVL1.
Recommended Dilutions: Western Blot: 1:500-1:1000; ELISA: 1:40000
Type: Primary
Antigen: ELOVL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse