Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-SLC27A4 Rabbit Polyclonal Antibody
  ABNOPAB17757
 :  Abnova
Anti-SLC27A4 Rabbit Polyclonal Antibody
  ABNOPAB17757
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 27 (fatty acid transporter), member 4
  • Antigen symbol:
    Solute carrier family 27, member 4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    10999
  • Antigen synonyms:
    ACSVL4|IPS|FATP4
  • Storage buffer:
    PBS, pH 7,4 (150mM NaCl, 0,02% sodium azide, 50% glycerol)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to internal of human SLC27A4.
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Rabbit polyclonal antibody raised against synthetic peptide of SLC27A4.

Recommended Dilutions: Immunofluorescence: 1:500-1:1000; ELISA: 1:40000

Type: Primary
Antigen: SLC27A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse