Anti-GABRG2 Goat Polyclonal Antibody
ABNOPAB18556
: Abnova
- Antibody type:Primary
- Antigen name:gamma-aminobutyric acid (GABA) A receptor, gamma 2
- Antigen symbol:GABRG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:internal
- Cross adsorption:No
- Format:Liquid
- Gene ID:2566
- Antigen synonyms:CAE2|ECA2|GEFSP3
- Amino acid number:400 to 410
- Storage buffer:0,5 mg/mL Tris saline, pH 7,3 (0,02% sodium azide, 0,5% BSA)
- Sequence:C-NNATHLQERDE
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 400-410 at internal region of human GABRG2.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Goat polyclonal antibody raised against synthetic peptide of GABRG2.
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3 ?g/ml
Type: Primary
Antigen: GABRG2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Mouse, Rat