Anti-PPAPDC1A Rabbit Polyclonal Antibody
Catalog # ABNOPAB19155
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Phosphatidic Acid Phosphatase Type 2 Domain-Containing Protein 1A
- Antigen symbol:PPAPDC1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:196051
- Antigen synonyms:Phosphatidate phosphatase PPAPDC1A|Phosphatidic acid phosphatase type 2 domain-containing protein 1A|PPAPDC1|PPAPDC1A|DPPL2
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 14 amino acids near C-terminus of human PPAPDC1A.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody rasied against synthetic peptide of PPAPDC1A.
Recommended Dilutions: Western Blot: 1-2 ?g/ml
Type: Primary
Antigen: PPAPDC1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat