Anti-SLC12A4 Goat Polyclonal Antibody
Catalog # ABNOPAB7375
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 12 (potassium/chloride transporter), member 4
- Antigen symbol:Solute carrier family 12, member A4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:6560
- Antigen synonyms:KCC1
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-DPSHAPDNFRE
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human SLC12A4.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of SLC12A4.
Recommended Dilutions: ELISA: 1:4000; Western Blot: 0.3-1 ?g/ml
Type: Primary
Antigen: SLC12A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human