Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-Gabra1 Rabbit Polyclonal Antibody
  ABNOPAB9680
 :  Abnova
Anti-Gabra1 Rabbit Polyclonal Antibody
  ABNOPAB9680
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    gamma-aminobutyric acid (GABA) A receptor, alpha 1
  • Antigen symbol:
    GABRA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Primates,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    29705
  • Storage buffer:
    10 mM HEPES, 150 mM NaCl, pH 7,5 (50% glycerol, 10% BSA)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to rat Gabra1.
  • Purification:
    Affinity purification
  • Size:
    100 μl
  • Pk:
    100 µg

 

 

Rabbit polyclonal antibody raised against a recombinant Gabra1.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: Gabra1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Primates, Rat