Anti-GABRB1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB9685
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Gamma-aminobutyric acid (GABA) A Receptor beta 1
- Antigen symbol:GABRB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Zebrafish,Primates,Chicken,Human,Bovine,Xenopus laevis,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:25450
- Antigen synonyms:GABA-RB|GABA(A) receptor|GABA(A)R beta-1|Gamma-aminobutyric-acid receptor beta-1 subunit precursor|GBRB1
- Storage buffer:10 mM HEPES, 150 mM NaCl, pH 7,5 (50% glycerol, 10% BSA)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to rat Gabrb1.
- Purification:Affinity purification
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against a recombinant Gabrb1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: GABRB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Clawed frog, Dog, Human, Mouse, Primates, Rat, Zebrafish